Search
Search

Acsl4| Gene Symbol: Acsl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033600.1 | NP_001028772.1 | 5, 6 | 817 | ||
| NM_019477.3 | NP_062350.3 | 5, 6 | 820 | |||
| NM_207625.2 | NP_997508.1 | 5, 6 | 1110 | |||
| GenBank mRNA | AB033885.1 | - | 5, 6 | 629 | ||
| AB033886.1 | - | 5, 6 | 919 | |||
| AB033887.1 | - | 5, 6 | 967 | |||
| AJ243502.1 | - | 5, 6 | 665 | |||
| AK054387.1 | - | 5, 6 | 790 | |||
| BC016416.1 | - | 5, 6 | 691 | 3157957 | ||
| BC058663.1 | - | 5, 6 | 906 | 6409748 | ||