Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KIR2DS3 SIRNA Assays ›
          Gene Symbol:
          KIR2DS3 KIR2DL1 KIR2DS1 KIR2DS2 KIR2DS4
          Gene Name:
          killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 3
          killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1
          killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 1
          killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 2
          killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4
          Gene Aliases:
          NKAT7
          CD158A, KIR-K64, KIR221, KIR2DS1, NKAT, NKAT-1, NKAT1, p58.1
          CD158H, CD158a, KIR2DP1DL1, KIR2DS4, p50.1
          183ActI, CD158J, CD158b, KIR-2DS2, NKAT-5, NKAT5, cl-49
          CD158I, KIR-2DS4, KIR1D, KIR2DS1, KIR412, KKA3, NKAT-8, NKAT8
          Chromosome Location:
          -
          Chromosome Location:
          Chr.19: 54769208 - 54784326 on Build GRCh38
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.19: 54832676 - 54848569 on Build GRCh38
          UniGene ID:
          Hs.744266
          Hs.654605
          Hs.744319
          Hs.512572
          Hs.654608
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID s7858

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KIR2DS3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012313.1 NP_036445.1 5, 6 720
          GenBank mRNA AF022047.1 - 5, 6 713
          AM851002.1 - 5, 6 745
          AY789063.2 - 4, 5 693
          L76670.1 - 5, 6 720
          X97231.1 - 5, 6 768
          Gene Symbol: KIR2DL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014218.2 NP_055033.2 5, 6 747
          XM_017026782.1 XP_016882271.1 5, 6 725
          GenBank mRNA AF285431.1 - 5, 6 742
          AF285432.1 - 4, 5 669
          AY366237.1 - 5, 6 745
          AY366238.1 - 5, 6 691
          AY366239.1 - 5, 6 739
          AY366241.1 - 5, 6 746
          AY789055.1 - 6, 7 742
          AY789056.1 - 5, 6 709
          KJ699235.1 - 5, 6 982
          L41267.1 - 6, 7 741
          U24076.1 - 6, 7 742
          Gene Symbol: KIR2DS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AF285437.1 - 4, 5 669
          Gene Symbol: KIR2DS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291695.1 NP_001278624.1 5, 6 744
          NM_001291696.1 NP_001278625.1 4, 5 444
          NM_001291700.1 NP_001278629.1 3, 4 150
          NM_012312.4 NP_036444.1 5, 6 744
          GenBank mRNA AF074087.1 - 3, 4 106
          AJ002102.1 - 5, 6 706
          AJ002103.1 - 5, 6 709
          AM409094.1 - 5, 6 710
          AY366243.1 - 5, 6 719
          AY366244.1 - 5, 6 708
          BC108917.1 - 5, 6 711 40017299
          BC119631.1 - 5, 6 728 40115568
          BC119632.1 - 5, 6 728 40115570
          BC125146.1 - 4, 5 693
          BC126856.1 - 5, 6 711 40017301
          FR750994.1 - 5, 6 744
          L41347.1 - 5, 6 728
          L76667.1 - 5, 6 426
          L76668.1 - 4, 5 420
          U24079.1 - 5, 6 707
          X89893.1 - 5, 6 709
          Gene Symbol: KIR2DS4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001281971.1 NP_001268900.1 5, 6 776
          NM_012314.5 NP_036446.3 6, 7 798
          GenBank mRNA AF002255.1 - 6, 7 755
          AJ417554.1 - 4, 5 667
          AJ417555.1 - 4, 5 689
          AJ620615.2 - 4, 5 660
          AJ620616.2 - 4, 5 660
          AY102624.1 - 3, 4 399
          AY366245.1 - 5, 6 743
          AY366246.1 - 5, 6 694
          AY388472.1 - 5, 6 747
          AY789064.2 - 5, 6 671
          BC096694.1 - 5, 6 720 40026979
          BC101977.1 - 5, 6 720 40004312
          BC103693.1 - 5, 6 720 40004314
          BC108918.1 - 6, 7 711 40017300
          BC125147.1 - 4, 5 693
          BC127084.1 - 5, 6 726 40120468
          BC144566.1 - 5, 6 798 9053094
          BC171788.1 - 5, 6 798 9054442
          L76671.1 - 5, 6 720
          U24077.1 - 5, 6 700
          X94609.1 - 5, 6 764

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular defense response
          natural killer cell inhibitory signaling pathway
          immune response
          regulation of immune response
          signal transduction
          innate immune response
          receptor activity
          protein binding
          transmembrane signaling receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0