Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KIR3DL1 SIRNA Assays ›
          Gene Symbol:
          KIR3DL1 KIR3DS1
          Gene Name:
          killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 1
          killer cell immunoglobulin like receptor, three Ig domains and short cytoplasmic tail 1
          Gene Aliases:
          CD158E1, KIR, KIR3DL1/S1, KIR3DL2, NKAT-3, NKAT3, NKB1, NKB1B
          CD158E2, KIR-123FM, KIR-G1, NKAT-10, NKAT10
          Chromosome Location:
          Chr.19: 54816438 - 54830778 on Build GRCh38
          Chromosome Location:
          -
          UniGene ID:
          Hs.645228
          Hs.743496
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s7867

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KIR3DL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001322168.1 NP_001309097.1 3 218
          NM_013289.2 NP_037421.2 3 218
          GenBank mRNA AF022049.1 - 3 155
          AF103010.1 - 3 155
          AF103011.1 - 3 155
          AF103012.1 - 3 155
          AF103013.1 - 3 155
          AF208688.1 - 1 92
          AF208689.1 - 1 92
          AF262968.1 - 1 74
          AF262971.1 - 1 74
          AF262972.1 - 1 74
          AF262973.1 - 3 188
          AJ417556.1 - 1 104
          AJ417557.1 - 1 104
          AY366236.1 - 3 193
          AY366249.1 - 3 155
          AY366250.1 - 3 181
          AY366251.1 - 3 155
          AY366252.1 - 3 155
          AY760050.1 - 3 155
          AY789065.1 - 3 170
          AY789066.1 - 3 155
          AY789072.1 - 3 155
          BC028206.1 - 3 182 5215674
          BC101540.1 - 3 174 8069046
          BC101542.1 - 3 174 8069048
          EF462885.1 - 1 85
          EF472674.1 - 1 85
          EF472675.1 - 1 85
          EF472676.1 - 1 85
          EF472677.1 - 1 85
          EF472678.1 - 1 85
          EF472679.1 - 1 85
          L41269.1 - 3 174
          U30273.1 - 3 157
          U30274.1 - 3 218
          U31416.1 - 3 180
          U33328.1 - 3 180
          X94262.1 - 3 156
          Gene Symbol: KIR3DS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001083539.2 NP_001077008.1 3 192
          GenBank mRNA AF022044.1 - 3 168
          AJ417558.3 - 1 104
          AK301140.1 - 3 192
          BC098288.1 - 3 166 40026980
          BC105679.1 - 3 166 40004313
          DC390157.1 - 3 192
          EF370406.1 - 2 116
          L76661.1 - 3 166
          U73396.1 - 3 155
          X97233.1 - 3 155

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          signal transduction
          natural killer cell mediated cytotoxicity
          regulation of immune response
          natural killer cell activation
          innate immune response
          HLA-B specific inhibitory MHC class I receptor activity
          MHC class I receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0