Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017395.2 | NP_059091.2 | 9 | 1683 | ||
GenBank mRNA | AB221679.1 | - | 9 | 1664 | ||
AF209854.1 | - | 9 | 1664 | |||
AK088686.1 | - | 9 | 1849 | |||
AK156195.1 | - | 1 | 307 | |||
BC051965.1 | - | 10 | 1720 | 5702640 | ||
BC057336.1 | - | 9 | 2073 | |||
BC075717.1 | - | 10 | 2156 | |||
BC118948.2 | - | 9 | 1664 | 40092396 | ||
BC118949.2 | - | 9 | 1664 | 40092397 |