Search
Search

Nagk| Gene Symbol: Nagk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164187.1 | NP_001157659.1 | 7 | 802 | ||
| NM_019542.2 | NP_062415.1 | 7 | 811 | |||
| GenBank mRNA | AJ242909.1 | - | 7 | 823 | ||
| AJ242911.1 | - | 7 | 763 | |||
| AK007240.1 | - | 7 | 806 | |||
| AK155292.1 | - | 7 | 703 | |||
| BC004689.1 | - | 7 | 689 | 3501470 | ||