Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Txnip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009935.2 | NP_001009935.1 | 5 | 1090 | ||
NM_023719.2 | NP_076208.2 | 5 | 1087 | |||
GenBank mRNA | AF173681.1 | - | 5 | 854 | ||
AF282826.1 | - | 5 | 808 | |||
AK004653.2 | - | 5 | 1087 | |||
AK082476.1 | - | 5 | 1085 | |||
AK088870.1 | - | 5 | 1085 | |||
AK089304.1 | - | 5 | 1090 | |||
AK089351.1 | - | 5 | 1088 | |||
AK089403.1 | - | 5 | 1090 | |||
AK155517.1 | - | 5 | 1088 | |||
AK171205.1 | - | 5 | 1088 | |||
AK171312.1 | - | 5 | 1088 | |||
AK171425.1 | - | 5 | 1088 | |||
AK171456.1 | - | 5 | 1085 | |||
AK171464.1 | - | 5 | 1088 | |||
AK171527.1 | - | 5 | 1088 | |||
AK200630.1 | - | 1 | 198 | |||
BC011212.1 | - | 5 | 1066 | 4193517 | ||
BC031850.1 | - | 5 | 1065 | 3591421 |