Search
Search

P2ry4| Gene Symbol: P2ry4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020621.4 | NP_065646.1 | 3 | 1255 | ||
| GenBank mRNA | AK076364.1 | - | 3 | 1256 | ||
| BC119206.1 | - | 2 | 1037 | 8733955 | ||
| BC120511.1 | - | 2 | 1037 | 40129434 | ||
Set Membership: |
|