Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RSAD2 SIRNA Assays ›
          Gene Symbol:
          Rsad2
          Gene Name:
          radical S-adenosyl methionine domain containing 2
          Gene Aliases:
          2510004L01Rik, Vig1, cig5
          Chromosome Location:
          Chr.12: 26442743 - 26456478 on Build GRCm38
          UniGene ID:
          Mm.24045
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s81520

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rsad2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021384.4 NP_067359.2 5, 6 960
          GenBank mRNA AF236064.2 - 5, 6 936
          AF442152.1 - 5, 6 921
          AK010906.1 - 5, 6 939
          AK036373.1 - 5, 6 940
          AK149900.1 - 5, 6 941
          AK150069.1 - 5, 6 942
          AK150347.1 - 5, 6 817
          AK150406.1 - 5, 6 940
          AK150425.1 - 5, 6 942
          AK150525.1 - 5, 6 941
          AK150536.1 - 5, 6 943
          AK150616.1 - 5, 6 940
          AK150684.1 - 5, 6 942
          AK150731.1 - 5, 6 942
          AK150767.1 - 5, 6 942
          AK150783.1 - 5, 6 942
          AK150788.1 - 5, 6 942
          AK150812.1 - 5, 6 942
          AK150835.1 - 5, 6 941
          AK151140.1 - 5, 6 942
          AK151185.1 - 5, 6 942
          AK151191.1 - 5, 6 942
          AK151206.1 - 5, 6 942
          AK151286.1 - 5, 6 940
          AK151308.1 - 5, 6 942
          AK151331.1 - 6, 7 1377
          AK151339.1 - 5, 6 940
          AK151389.1 - 5, 6 942
          AK151410.1 - 5, 6 941
          AK151412.1 - 5, 6 940
          AK151414.1 - 5, 6 941
          AK151476.1 - 5, 6 942
          AK151502.1 - 5, 6 942
          AK151538.1 - 5, 6 942
          AK151767.1 - 5, 6 942
          AK151770.1 - 5, 6 940
          AK151825.1 - 5, 6 940
          AK151833.1 - 5, 6 940
          AK151846.1 - 5, 6 940
          AK151906.1 - 5, 6 942
          AK151909.1 - 5, 6 940
          AK151940.1 - 5, 6 940
          AK151954.1 - 5, 6 942
          AK151971.1 - 5, 6 942
          AK152050.1 - 5, 6 940
          AK152065.1 - 5, 6 942
          AK152075.1 - 5, 6 942
          AK152235.1 - 5, 6 940
          AK152239.1 - 5, 6 942
          AK152241.1 - 5, 6 942
          AK152310.1 - 5, 6 940
          AK152354.1 - 5, 6 940
          AK152493.1 - 5, 6 942
          AK152600.1 - 5, 6 940
          AK152601.1 - 5, 6 942
          AK152631.1 - 5, 6 940
          AK152644.1 - 5, 6 942
          AK152664.1 - 5, 6 942
          AK152701.1 - 5, 6 940
          AK152796.1 - 5, 6 940
          AK152861.1 - 5, 6 940
          AK152880.1 - 5, 6 940
          AK152895.1 - 5, 6 942
          AK152934.1 - 5, 6 942
          AK152962.1 - 5, 6 940
          AK153003.1 - 5, 6 942
          AK153024.1 - 5, 6 942
          AK153062.1 - 5, 6 944
          AK153077.1 - 5, 6 940
          AK153078.1 - 5, 6 940
          AK153232.1 - 5, 6 942
          AK153234.1 - 5, 6 942
          AK153306.1 - 5, 6 940
          AK153318.1 - 5, 6 942
          AK153324.1 - 5, 6 942
          AK153350.1 - 6, 7 1377
          AK153435.1 - 5, 6 818
          AK153453.1 - 5, 6 906
          AK153556.1 - 5, 6 942
          AK159147.1 - 5, 6 940
          BC057868.1 - 5, 6 933 5327504
          CT010378.1 - 5, 6 927

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_080657, AK312670, BC017969, AK300085, AF442151.

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          immune system process
          response to virus
          regulation of ossification
          positive regulation of toll-like receptor 7 signaling pathway
          positive regulation of toll-like receptor 9 signaling pathway
          CD4-positive, alpha-beta T cell activation
          CD4-positive, alpha-beta T cell differentiation
          negative regulation of viral genome replication
          innate immune response
          negative regulation of protein secretion
          defense response to virus
          positive regulation of T-helper 2 cell cytokine production
          catalytic activity
          protein binding
          protein self-association
          metal ion binding
          iron-sulfur cluster binding
          4 iron, 4 sulfur cluster binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0