Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LGALS9 SIRNA Assays ›
          Gene Symbol:
          LGALS9
          Gene Name:
          galectin 9
          Gene Aliases:
          HUAT, LGALS9A
          Chromosome Location:
          Chr.17: 27631148 - 27649560 on Build GRCh38
          UniGene ID:
          Hs.81337
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s8161

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LGALS9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002308.3 NP_002299.2 9 866
          NM_009587.2 NP_033665.1 10 962
          NR_024043.1 9 795
          XM_006721892.2 XP_006721955.1 8 793
          XM_017024623.1 XP_016880112.1 9 918
          GenBank mRNA AB003517.1 - 9 773
          AB005894.1 - 9 807
          AB006782.1 - 10 925
          AB008492.1 - 10 869
          AK126017.1 - 9 2216
          AK223232.1 - 9 830
          AK296031.1 - 9 737
          BC105942.1 - 9 790 40031795
          BC105944.1 - 9 790 40031802
          BC110340.1 - 9 807 5760999
          Z49107.1 - 9 835

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          natural killer cell tolerance induction
          chemotaxis
          inflammatory response
          female pregnancy
          positive regulation of gene expression
          negative regulation of gene expression
          response to lipopolysaccharide
          regulation of interleukin-4 production
          regulation of interleukin-5 production
          negative regulation of chemokine production
          negative regulation of interferon-gamma production
          negative regulation of tumor necrosis factor production
          positive regulation of interleukin-4 production
          positive regulation of CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation involved in immune response
          toll-like receptor 2 signaling pathway
          toll-like receptor 4 signaling pathway
          p38MAPK cascade
          positive regulation of NF-kappaB import into nucleus
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of mast cell degranulation
          negative regulation of natural killer cell mediated cytotoxicity
          negative regulation of activated T cell proliferation
          positive regulation of viral entry into host cell
          positive regulation of interleukin-1 beta secretion
          positive regulation of NF-kappaB transcription factor activity
          maternal process involved in female pregnancy
          positive regulation of activated T cell autonomous cell death
          ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          response to interleukin-1
          cellular response to interferon-gamma
          positive regulation of transforming growth factor beta production
          positive regulation of monocyte chemotactic protein-1 production
          mature conventional dendritic cell differentiation
          cellular response to virus
          regulation of p38MAPK cascade
          positive regulation of interferon-gamma secretion
          positive regulation of tumor necrosis factor secretion
          positive regulation of interleukin-8 secretion
          positive regulation of dendritic cell chemotaxis
          negative regulation of CD4-positive, alpha-beta T cell proliferation
          positive regulation of CD4-positive, alpha-beta T cell proliferation
          positive regulation of interleukin-13 secretion
          positive regulation of dendritic cell apoptotic process
          positive regulation of interleukin-6 secretion
          positive regulation of interleukin-10 secretion
          positive regulation of interleukin-12 secretion
          positive regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
          positive regulation of dendritic cell differentiation
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          signal transducer activity
          galactose binding
          enzyme binding
          carbohydrate binding
          disaccharide binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0