Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TPM3 SIRNA Assays ›
          Gene Symbol:
          Tpm3
          Gene Name:
          tropomyosin 3, gamma
          Gene Aliases:
          TM30nm, TMnm, Tm5NM, Tpm-5, Tpm5, Trop-5, gamma-TM, hTM30nm, hTMnm
          Chromosome Location:
          Chr.3: 90072651 - 90100902 on Build GRCm38
          UniGene ID:
          Mm.240839
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s81894

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tpm3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001253738.1 NP_001240667.1 4 628
          NM_001253740.1 NP_001240669.1 4 628
          NM_001271764.1 NP_001258693.1 4 628
          NM_001293748.1 NP_001280677.2 5 611
          NM_001293749.1 NP_001280678.1 4 628
          NM_022314.3 NP_071709.3 5 611
          GenBank mRNA AF317223.1 - 5 525
          AK050665.1 - 4 474
          AK088111.1 - 4 488
          AK088902.1 - 4 486
          AK145238.1 - 4 485
          AK145243.1 - 4 485
          AK145804.1 - 4 485
          AK146349.1 - 4 458
          AK146557.1 - 4 610
          AK151441.1 - 4 510
          AK151500.1 - 4 485
          AK151571.1 - 4 484
          AK151578.1 - 4 485
          AK151595.1 - 4 457
          AK151602.1 - 4 485
          AK151991.1 - 4 484
          AK152260.1 - 4 484
          AK152659.1 - 4 457
          AK153031.1 - 4 457
          AK160011.1 - 4 510
          AK164086.1 - 4 485
          AK165428.1 - 4 484
          AK166836.1 - 4 472
          AK167268.1 - 4 462
          AK167534.1 - 4 490
          AK167581.1 - 4 485
          AK168843.1 - 4 453
          AK169102.1 - 4 456
          AK169245.1 - 4 461
          AK169821.1 - 4 502
          BC029186.1 - 5 568 4502880
          BC055407.1 - 3754
          BC092045.1 - 4 461 3591372
          CX237327.1 - 4 450
          U04541.1 - 5 581
          X53753.1 - 4 496

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001278190, NM_001043351, NM_001043353, NM_001043352, NM_001278189, and others.
          This siRNA also targets the following transcripts in Rat: NM_001301285, NM_173111, NM_001301286, NM_057208, XM_006232580, and others.

          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Process(es)

          actin binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0