Search
Search

Dctn5| Gene Symbol: Dctn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021608.3 | NP_067621.3 | 3, 4 | 331 | ||
| GenBank mRNA | AF190795.1 | - | 3, 4 | 302 | ||
| AK010238.1 | - | 3, 4 | 311 | |||
| AK051957.1 | - | 3, 4 | 270 | |||
| AK076100.1 | - | 3, 4 | 304 | |||
| AK076738.1 | - | 3, 4 | 331 | |||
| AK145958.1 | - | 2, 3 | 1282 | |||
| AK146123.1 | - | 3, 4 | 297 | |||
| AK151193.1 | - | 3, 4 | 303 | |||
| AK151876.1 | - | 3, 4 | 303 | |||
| AK168983.1 | - | 3, 4 | 311 | |||
| BC005671.1 | - | 2, 3 | 1305 | 3153175 | ||
| BC010834.1 | - | 3, 4 | 300 | 4235312 | ||
| BC092273.1 | - | 2, 3 | 1284 | |||