Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCL21C SIRNA Assays ›
          Gene Symbol:
          Ccl21c Ccl21a Ccl21b Gm10591 Gm13304 Gm1987 Gm21541
          Gene Name:
          chemokine (C-C motif) ligand 21C (leucine)
          chemokine (C-C motif) ligand 21A (serine)
          chemokine (C-C motif) ligand 21B (leucine)
          predicted gene 10591
          predicted gene 13304
          predicted gene 1987
          predicted gene, 21541
          Gene Aliases:
          6Ckine, CKb9, Ccl21b, SLC, Scya21, Scya21b, Scya21c, TCA4, exodus-2
          6CKBAC2, 6Ckine, ALP, AW987545, CKb9, SCYA21a, SLC, Scya21, Scya21b, Tca4, plt
          6CKBAC1, 6Ckine, AI159701, CKb9, SLC, Scya21, Scya21a, TCA4
          ENSMUSG00000073869
          ENSMUSG00000073878, OTTMUSG00000011350
          100038965
          -
          Chromosome Location:
          Chr.4|NT_187053: 59769 - 60899 on Build GRCm38
          Chromosome Location:
          Chr.4: 42772860 - 42773991 on Build GRCm38
          Chromosome Location:
          Chr.4: 42255731 - 42256825 on Build GRCm38
          Chromosome Location:
          Chr.4: 42612123 - 42613253 on Build GRCm38
          Chromosome Location:
          Chr.4: 42114787 - 42115917 on Build GRCm38
          Chromosome Location:
          Chr.4|NT_187054: 120655 - 121788 on Build GRCm38
          Chromosome Location:
          Chr.4: 41903613 - 41904743 on Build GRCm38
          UniGene ID:
          Mm.407493
          Mm.220853
          Mm.407493
          Mm.407493
          Mm.407493
          Mm.220853
          Mm.407493
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s82413

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ccl21c
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_023052.2 NP_075539.1 4 837
          GenBank mRNA BC087957.1 - 4 837 6750979
          U88322.1 - 4 810
          Gene Symbol: Ccl21a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011124.4 NP_035254.1 4 838
          GenBank mRNA AF001980.1 - 4 766
          AK144258.1 - 4 840
          AK155924.1 - 4 838
          BC025974.1 - 4 837 4208773
          BC028747.1 - 4 825 1196234
          BC038120.1 - 4 807 3372243
          Gene Symbol: Ccl21b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011335.2 NP_035465.2 801
          Gene Symbol: Gm10591
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001193668.1 NP_001180597.1 837
          Gene Symbol: Gm13304
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001193666.1 NP_001180595.1 837
          Gene Symbol: Gm1987
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001193667.1 NP_001180596.1 4 840
          Gene Symbol: Gm21541
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270360.1 NP_001257289.1 4 837
          GenBank mRNA AA123440.1 - 1 371

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_002989, AB002409, BC027918, U88320, AF001979, and others.

          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          cell chemotaxis
          lymph node development
          induction of positive chemotaxis
          positive regulation of T cell migration
          leukocyte chemotaxis
          negative regulation of myeloid cell differentiation
          biological_process
          chemokine activity
          chemokine receptor binding
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0