Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASPH SIRNA Assays ›
Gene Symbol:
Asph
Gene Name:
aspartate-beta-hydroxylase
Gene Aliases:
2310005F16Rik, 3110001L23Rik, AI848629, AW261690, AW561901, BAH, C79816, cI-37
Chromosome Location:
Chr.4: 9449085 - 9669344 on Build GRCm38
UniGene ID:
Mm.222206
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s82444
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Asph
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001177849.1
NP_001171320.1
2
410
NM_001177850.1
NP_001171321.1
2
410
NM_001177851.1
NP_001171322.1
2
410
NM_001177852.1
NP_001171323.1
2
211
NM_001177853.1
NP_001171324.1
2
211
NM_001177854.1
NP_001171325.1
2
211
NM_001177855.1
NP_001171326.1
2
410
NM_001177856.1
NP_001171327.1
2
410
NM_001290367.1
NP_001277296.1
2
217
NM_023066.3
NP_075553.2
2
410
NM_133723.2
NP_598484.2
2
211
GenBank mRNA
AF221854.1
-
2
186
AF223413.1
-
2
211
AF223414.1
-
2
221
AF289486.1
-
2
332
AF289487.1
-
2
332
AF289488.1
-
2
332
AF289490.1
-
2
168
AF289491.1
-
2
168
AF289492.1
-
2
168
AF302653.2
-
2
110
AF302654.1
-
2
163
AF302655.1
-
2
90
AK009170.1
-
2
136
AK009903.1
-
2
147
AK014674.1
-
2
144
AK017453.1
-
2
143
AK028679.1
-
2
142
AK030293.1
-
2
334
AK030694.1
-
2
145
AK032420.1
-
2
333
AK035735.1
-
2
219
AK049506.1
-
2
334
AK151787.1
-
2
333
AK160984.1
-
2
335
AK165260.1
-
2
356
BC015281.1
-
2
319
BC061098.1
-
2
356
BC128299.1
-
2
139
BC152365.1
-
2
139
BY268464.1
-
2
412
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
pattern specification process
negative regulation of cell proliferation
positive regulation of calcium ion transport into cytosol
peptidyl-amino acid modification
regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
regulation of protein stability
activation of store-operated calcium channel activity
response to ATP
limb morphogenesis
peptidyl-aspartic acid hydroxylation
positive regulation of proteolysis
positive regulation of transcription, DNA-templated
regulation of cytosolic calcium ion concentration
oxidation-reduction process
palate development
face morphogenesis
calcium ion transmembrane transport
cellular response to calcium ion
positive regulation of intracellular protein transport
activation of cysteine-type endopeptidase activity
regulation of protein depolymerization
peptide-aspartate beta-dioxygenase activity
calcium ion binding
protein binding
oxidoreductase activity
metal ion binding
dioxygenase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0