Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ASPH SIRNA Assays ›
          Gene Symbol:
          Asph
          Gene Name:
          aspartate-beta-hydroxylase
          Gene Aliases:
          2310005F16Rik, 3110001L23Rik, AI848629, AW261690, AW561901, BAH, C79816, cI-37
          Chromosome Location:
          Chr.4: 9449085 - 9669344 on Build GRCm38
          UniGene ID:
          Mm.222206
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s82445

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Asph
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177849.1 NP_001171320.1 2, 3 530
          NM_001177850.1 NP_001171321.1 2, 3 530
          NM_001177851.1 NP_001171322.1 2, 3 530
          NM_001177852.1 NP_001171323.1 2, 3 331
          NM_001177853.1 NP_001171324.1 2, 3 331
          NM_001177854.1 NP_001171325.1 2, 3 331
          NM_001177855.1 NP_001171326.1 2, 3 530
          NM_001290367.1 NP_001277296.1 2, 3 337
          NM_023066.3 NP_075553.2 2, 3 530
          NM_133723.2 NP_598484.2 2, 3 331
          GenBank mRNA AF221854.1 - 2, 3 306
          AF223413.1 - 2, 3 331
          AF223414.1 - 2, 3 341
          AF289486.1 - 2, 3 452
          AF289487.1 - 2, 3 452
          AF289488.1 - 2, 3 452
          AF289490.1 - 2, 3 288
          AF289491.1 - 2, 3 288
          AF289492.1 - 2, 3 288
          AF302653.2 - 2, 3 230
          AF302654.1 - 2, 3 283
          AF302655.1 - 2, 3 210
          AK009170.1 - 2, 3 256
          AK009903.1 - 2, 3 267
          AK014674.1 - 2, 3 264
          AK017453.1 - 2, 3 263
          AK028679.1 - 2, 3 262
          AK030293.1 - 2, 3 454
          AK030694.1 - 2, 3 265
          AK035735.1 - 2, 3 339
          AK049506.1 - 2, 3 454
          AK151787.1 - 2, 3 453
          AK160984.1 - 2, 3 455
          AK165260.1 - 2, 3 476
          BC015281.1 - 2, 3 439
          BC128299.1 - 2, 3 259
          BC152365.1 - 2, 3 259

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          pattern specification process
          negative regulation of cell proliferation
          positive regulation of calcium ion transport into cytosol
          peptidyl-amino acid modification
          regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
          regulation of protein stability
          activation of store-operated calcium channel activity
          response to ATP
          limb morphogenesis
          peptidyl-aspartic acid hydroxylation
          positive regulation of proteolysis
          positive regulation of transcription, DNA-templated
          regulation of cytosolic calcium ion concentration
          oxidation-reduction process
          palate development
          face morphogenesis
          calcium ion transmembrane transport
          cellular response to calcium ion
          positive regulation of intracellular protein transport
          activation of cysteine-type endopeptidase activity
          regulation of protein depolymerization
          peptide-aspartate beta-dioxygenase activity
          calcium ion binding
          protein binding
          oxidoreductase activity
          metal ion binding
          dioxygenase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0