Search
Search

Crnkl1| Gene Symbol: Crnkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025820.3 | NP_080096.1 | 8 | 1088 | ||
| GenBank mRNA | AK004749.1 | - | 8 | 1089 | ||
| AK012962.1 | - | 8 | 1099 | |||
| AK020002.1 | - | 8 | 1105 | |||
| AK088882.1 | - | 8 | 1092 | |||
| BC029187.1 | - | 8 | 1053 | 3495255 | ||