Search
Search

Mtap| Gene Symbol: Mtap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024433.2 | NP_077753.1 | 7 | 876 | ||
| GenBank mRNA | AB056100.1 | - | 7 | 864 | ||
| AK005064.1 | - | 7 | 823 | |||
| AK011421.1 | - | 7 | 821 | |||
| AK089916.1 | - | 7 | 813 | |||
| AK167319.1 | - | 7 | 877 | |||
| BC003858.1 | - | 7 | 820 | 3485422 | ||