Search
Search

Prpf18| Gene Symbol: Prpf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026045.3 | NP_080321.2 | 9 | 1013 | ||
| GenBank mRNA | AK028921.1 | - | 2 | 150 | ||
| AK035073.1 | - | 9 | 1043 | |||
| AK135707.1 | - | 9 | 1053 | |||
| AK135890.1 | - | 9 | 1034 | |||
| AK139720.1 | - | 9 | 1001 | |||
| AK161550.1 | - | 5 | 457 | |||
| BC004573.1 | - | 9 | 1013 | 3586127 | ||
| BC028306.1 | - | 9 | 1016 | |||
| BC058631.1 | - | 9 | 1008 | 6402639 | ||