Search
Search

Ttc33| Gene Symbol: Ttc33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026213.3 | NP_080489.1 | 5 | 675 | ||
| GenBank mRNA | AK010758.1 | - | 5 | 494 | ||
| AK013464.1 | - | 5 | 521 | |||
| AK050426.1 | - | 5 | 515 | |||
| AK075643.1 | - | 5 | 501 | |||
| BC017533.1 | - | 4 | 440 | |||
| BC061018.1 | - | 5 | 675 | 6590550 | ||