Search
Search

Rnf125| Gene Symbol: Rnf125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026301.2 | NP_080577.1 | 3 | 340 | ||
| GenBank mRNA | AB259692.1 | - | 3 | 371 | ||
| AK006569.1 | - | 2 | 168 | |||
| AK016103.1 | - | 3 | 537 | |||
| AK050395.1 | - | 3 | 341 | |||
| BC094290.1 | - | 2 | 137 | |||
| BC116883.1 | - | 2 | 209 | 40126202 | ||
| BC116887.1 | - | 2 | 209 | 40126206 | ||