Search
Search

2310057J18Rik
2310065H11Rik| Gene Symbol: 2310057J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026336.3 | NP_080612.1 | 3 | 517 | ||
| GenBank mRNA | AK008407.1 | - | 3 | 515 | ||
| AK008932.1 | - | 3 | 644 | |||
| AK009183.1 | - | 3 | 516 | |||
| AK009278.1 | - | 3 | 515 | |||
| AK009369.1 | - | 3 | 514 | |||
| AK009472.1 | - | 3 | 515 | |||
| AK009516.1 | - | 3 | 515 | |||
| AK009594.1 | - | 3 | 515 | |||
| AK009610.1 | - | 3 | 515 | |||
| AK009872.1 | - | 3 | 516 | |||
| AK009911.1 | - | 3 | 516 | |||
| AK009921.1 | - | 3 | 515 | |||
| AK009961.1 | - | 3 | 518 | |||
| AK010028.1 | - | 3 | 515 | |||
| AK010037.1 | - | 3 | 518 | |||
| AK010047.1 | - | 3 | 513 | |||
| AK010108.1 | - | 3 | 515 | |||
| AK010222.1 | - | 3 | 517 | |||
| AK010978.1 | - | 3 | 518 | |||
| AK075881.1 | - | 3 | 514 | |||
| AK075892.1 | - | 2 | 204 | |||
| AK075901.1 | - | 3 | 226 | |||
| BC021783.1 | - | 3 | 502 | 4458723 | ||
| BC055432.1 | - | 3 | 504 | |||
| Gene Symbol: 2310065H11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK010046.1 | - | 3 | 515 | ||