Search
Search

2310057J18Rik
2310065H11Rik| Gene Symbol: 2310057J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026336.3 | NP_080612.1 | 5 | 828 | ||
| GenBank mRNA | AK008407.1 | - | 5 | 826 | ||
| AK008932.1 | - | 5 | 950 | |||
| AK009183.1 | - | 5 | 827 | |||
| AK009278.1 | - | 5 | 826 | |||
| AK009369.1 | - | 5 | 825 | |||
| AK009472.1 | - | 5 | 826 | |||
| AK009516.1 | - | 5 | 826 | |||
| AK009574.1 | - | 5 | 826 | |||
| AK009594.1 | - | 5 | 826 | |||
| AK009872.1 | - | 5 | 827 | |||
| AK009911.1 | - | 5 | 827 | |||
| AK009921.1 | - | 5 | 826 | |||
| AK009961.1 | - | 5 | 830 | |||
| AK010028.1 | - | 5 | 826 | |||
| AK010037.1 | - | 5 | 829 | |||
| AK010047.1 | - | 5 | 824 | |||
| AK010108.1 | - | 5 | 826 | |||
| AK010222.1 | - | 5 | 828 | |||
| AK010978.1 | - | 5 | 830 | |||
| AK075881.1 | - | 5 | 825 | |||
| AK075892.1 | - | 4 | 515 | |||
| AK075901.1 | - | 5 | 537 | |||
| BC021783.1 | - | 5 | 813 | 4458723 | ||
| Gene Symbol: 2310065H11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK010046.1 | - | 5 | 827 | ||