Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MBNL1 SIRNA Assays ›
          Gene Symbol:
          MBNL1
          Gene Name:
          muscleblind like splicing regulator 1
          Gene Aliases:
          EXP, MBNL
          Chromosome Location:
          Chr.3: 152243656 - 152465780 on Build GRCh38
          UniGene ID:
          Hs.201858
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s8554

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MBNL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001314057.1 NP_001300986.1 11 1788
          NM_021038.4 NP_066368.2 9 2519
          NM_207292.2 NP_997175.1 8 2483
          NM_207293.1 NP_997176.1 8 1912
          NM_207294.1 NP_997177.1 6 1654
          NM_207295.1 NP_997178.1 7 1690
          NM_207296.1 NP_997179.1 7 1768
          NM_207297.1 NP_997180.1 7 1799
          XM_005247457.4 XP_005247514.1 11 2141
          XM_005247458.4 XP_005247515.1 11 3395
          XM_005247460.4 XP_005247517.1 11 2104
          XM_005247461.4 XP_005247518.1 10 2105
          XM_005247462.4 XP_005247519.1 10 2087
          XM_005247463.4 XP_005247520.1 9 2051
          XM_005247464.4 XP_005247521.1 10 2199
          XM_005247465.4 XP_005247522.1 10 2046
          XM_005247466.4 XP_005247523.1 9 2140
          XM_005247467.4 XP_005247524.1 9 2010
          XM_005247468.4 XP_005247525.1 8 1956
          XM_005247471.4 XP_005247528.1 8 2022
          XM_005247474.3 XP_005247531.1 9 1059
          XM_005247475.3 XP_005247532.1 10 1382
          XM_005247476.4 XP_005247533.1 8 1095
          XM_005247477.4 XP_005247534.1 9 1148
          XM_006713639.1 XP_006713702.1 11 2123
          XM_011512843.2 XP_011511145.1 10 1937
          XM_011512845.2 XP_011511147.1 9 1883
          XM_011512846.2 XP_011511148.1 8 1847
          XM_011512847.2 XP_011511149.1 8 1846
          XM_011512848.2 XP_011511150.1 11 1745
          XM_011512849.2 XP_011511151.1 7 1754
          XM_017006423.1 XP_016861912.1 10 2392
          XM_017006424.1 XP_016861913.1 10 2219
          XM_017006425.1 XP_016861914.1 9 2185
          XM_017006426.1 XP_016861915.1 9 2183
          XM_017006427.1 XP_016861916.1 9 2312
          XM_017006428.1 XP_016861917.1 9 1976
          XM_017006429.1 XP_016861918.1 9 2059
          XM_017006430.1 XP_016861919.1 8 2087
          XM_017006431.1 XP_016861920.1 8 2164
          XM_017006432.1 XP_016861921.1 9 2081
          XM_017006433.1 XP_016861922.1 8 2045
          XM_017006434.1 XP_016861923.1 8 2051
          XM_017006435.1 XP_016861924.1 10 1189
          XM_017006436.1 XP_016861925.1 8 3153
          XM_017006437.1 XP_016861926.1 7 2172
          XM_017006438.1 XP_016861927.1 8 1808
          XM_017006439.1 XP_016861928.1 9 1328
          XM_017006440.1 XP_016861929.1 8 1062
          XM_017006441.1 XP_016861930.1 7 1059
          XM_017006442.1 XP_016861931.1 7 1162
          XM_017006443.1 XP_016861932.1 6 1075
          GenBank mRNA AB007888.1 - 8 2483
          AF255334.1 - 6 1376
          AF395876.1 - 7 901
          AF401998.1 - 8 1105
          AF497718.1 - 7 982
          AF497719.1 - 7 1010
          AJ308400.1 - 8 1134
          AK297133.1 - 5 574
          AK304089.1 - 8 868
          AK307420.1 - 6 614
          BC043493.1 - 9 2024 6044671
          BC050535.1 - 11 1309 6042311
          BX537491.1 - 1 1592
          CR749454.1 - 3 147
          Y13829.1 - 8 1634

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001191566, XM_008761040, XM_006232430, XM_008761037, XM_008761042, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001253710, NM_001253711, NM_001253713, NM_001253708, NM_020007, and others.

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          mRNA processing
          nervous system development
          RNA splicing
          embryonic limb morphogenesis
          regulation of RNA splicing
          myoblast differentiation
          regulatory region RNA binding
          RNA binding
          double-stranded RNA binding
          protein binding
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0