Search
Search

| Gene Symbol: Fdx1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039824.2 | NP_001034913.1 | 5 | 732 | ||
| GenBank mRNA | AK008401.2 | - | 5 | 730 | ||
| AK010211.1 | - | 5 | 703 | |||
| AK020979.1 | - | 5 | 736 | |||
| BC061189.1 | - | 4 | 819 | |||
| Gene Symbol: Gm38431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004919.1 | - | 17 | 2936 | ||
| AY275472.1 | - | 17 | 2922 | |||