Search
Search

Fam198b| Gene Symbol: Fam198b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133187.3 | NP_573450.2 | 1 | 1003 | ||
| GenBank mRNA | AF285091.1 | - | 1 | 945 | ||
| AK053058.1 | - | 1 | 877 | |||
| AK143522.1 | - | 1 | 734 | |||
| BC056975.1 | - | 1 | 772 | 6827227 | ||
| EU797522.1 | - | 1 | 781 | |||