Search
Search

MPP1| Gene Symbol: MPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166460.1 | NP_001159932.1 | 10 | 1205 | ||
| NM_001166461.1 | NP_001159933.1 | 10 | 1196 | |||
| NM_001166462.1 | NP_001159934.1 | 11 | 1405 | |||
| NM_002436.3 | NP_002427.1 | 10 | 1256 | |||
| XM_011531167.1 | XP_011529469.1 | 9 | 1169 | |||
| XM_011531169.1 | XP_011529471.1 | 6 | 828 | |||
| GenBank mRNA | AK093702.1 | - | 1 | 1588 | ||
| AK223396.1 | - | 10 | 1236 | |||
| AK290246.1 | - | 10 | 1151 | |||
| AK303111.1 | - | 10 | 1071 | |||
| AK304538.1 | - | 10 | 1163 | |||
| AK311541.1 | - | 11 | 1136 | |||
| AK312296.1 | - | 10 | 1127 | |||
| AK315957.1 | - | 11 | 1254 | |||
| AY423731.1 | - | 10 | 1128 | |||
| AY634686.1 | - | 10 | 1066 | |||
| BC002392.2 | - | 10 | 1132 | 2820598 | ||
| M64925.1 | - | 10 | 1205 | |||