Search
Search

5430427O19Rik| Gene Symbol: 5430427O19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163539.1 | NP_001157011.1 | 2 | 211 | ||
| GenBank mRNA | AK017351.1 | - | 2 | 153 | ||
| AK155591.1 | - | 2 | 137 | |||
| BC115576.1 | - | 2 | 112 | 40102201 | ||
| BC115577.1 | - | 2 | 112 | 40102202 | ||
| BY203970.1 | - | 2 | 210 | |||