Search
Search

Armcx3| Gene Symbol: Armcx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027870.3 | NP_082146.2 | 5 | 1222 | ||
| GenBank mRNA | AK004598.1 | - | 5 | 1193 | ||
| AK030729.1 | - | 5 | 1195 | |||
| AK148876.1 | - | 1 | 683 | |||
| BC011101.1 | - | 1 | 729 | 3490748 | ||
| BC051113.1 | - | 5 | 1183 | 5249601 | ||