Search
Search

Armcx3| Gene Symbol: Armcx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027870.3 | NP_082146.2 | 5 | 738 | ||
| GenBank mRNA | AK004598.1 | - | 5 | 709 | ||
| AK030729.1 | - | 5 | 711 | |||
| AK148876.1 | - | 1 | 199 | |||
| BC011101.1 | - | 1 | 245 | 3490748 | ||
| BC051113.1 | - | 5 | 699 | 5249601 | ||
| BC108360.1 | - | 5 | 679 | |||
| BU505969.1 | - | 5 | 613 | |||