Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS6KB1 SIRNA Assays ›
          Gene Symbol:
          Rps6kb1
          Gene Name:
          ribosomal protein S6 kinase, polypeptide 1
          Gene Aliases:
          2610318I15Rik, 4732464A07Rik, 70kDa, AA959758, AI256796, AI314060, S6K1, p70/85s6k, p70s6k
          Chromosome Location:
          Chr.11: 86498869 - 86544807 on Build GRCm38
          UniGene ID:
          Mm.394280
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s91055

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps6kb1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001114334.1 NP_001107806.1 6 647
          NM_028259.4 NP_082535.1 6 647
          GenBank mRNA AK012045.1 - 1 26
          AK032724.1 - 6 609
          AK085687.1 - 6 616
          AK135716.1 - 6 615
          AK163258.1 - 6 617
          BC038491.1 - 6 620 4010710

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001272042, NM_001272044, NM_003161, NM_001272043, NM_001272060, and others.
          This siRNA also targets the following transcripts in Rat: NM_031985, XM_008768121, XM_008768120, XM_006247111, M58340, and others.

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          behavioral fear response
          regulation of translation
          protein phosphorylation
          apoptotic process
          cell cycle
          signal transduction
          germ cell development
          positive regulation of smooth muscle cell migration
          phosphorylation
          TOR signaling
          response to insulin
          cellular response to insulin stimulus
          response to tumor necrosis factor
          negative regulation of apoptotic process
          protein kinase B signaling
          positive regulation of translation
          positive regulation of mitotic cell cycle
          positive regulation of translational initiation
          regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          positive regulation of skeletal muscle tissue growth
          positive regulation of smooth muscle cell proliferation
          cellular response to growth factor stimulus
          cellular response to organic cyclic compound
          negative regulation of extrinsic apoptotic signaling pathway
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          ribosomal protein S6 kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          PDZ domain binding
          peptide binding
          protein phosphatase 2A binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0