Search
Search

Kcmf1| Gene Symbol: Kcmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019715.2 | NP_062689.2 | 2 | 486 | ||
| GenBank mRNA | AK007070.1 | - | 2 | 433 | ||
| AK142436.1 | - | 2 | 243 | |||
| AK183540.1 | - | 1 | 20 | |||
| BC029325.1 | - | 2 | 308 | |||
| BC043330.1 | - | 2 | 267 | 5249829 | ||