Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NBN SIRNA Assays ›
          Gene Symbol:
          NBN
          Gene Name:
          nibrin
          Gene Aliases:
          AT-V1, AT-V2, ATV, NBS, NBS1, P95
          Chromosome Location:
          Chr.8: 89933336 - 89984733 on Build GRCh38
          UniGene ID:
          Hs.492208
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s9292

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NBN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001024688.2 NP_001019859.1 4 429
          NM_002485.4 NP_002476.2 3 326
          XM_011517045.1 XP_011515347.1 4 259
          XM_011517046.1 XP_011515348.1 3 353
          XM_017013460.1 XP_016868949.1 4 320
          XM_017013461.1 XP_016868950.1 3 253
          XM_017013462.1 XP_016868951.1 4 438
          GenBank mRNA AF051334.1 - 3 267
          AF058696.2 - 3 268
          AK001017.1 - 3 322
          AK223256.1 - 3 270
          AK289848.1 - 3 322
          AK312410.1 - 3 328
          BC005293.1 - 3 248
          BC016762.1 - 3 240
          BC108650.1 - 4 268
          BC136802.1 - 3 285 9020792
          BC136803.1 - 3 285 9020793
          BC144413.1 - 3 285
          BC146797.1 - 3 278 8860301
          BX640816.1 - 4 376
          DA634817.1 - 4 429

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          damaged DNA-binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage checkpoint
          telomere maintenance
          double-strand break repair via homologous recombination
          DNA double-strand break processing
          DNA synthesis involved in DNA repair
          strand displacement
          blastocyst growth
          DNA replication
          double-strand break repair
          double-strand break repair via nonhomologous end joining
          cell cycle arrest
          mitotic cell cycle checkpoint
          mitotic G2 DNA damage checkpoint
          cell proliferation
          regulation of DNA-dependent DNA replication initiation
          DNA damage response, signal transduction by p53 class mediator
          telomeric 3' overhang formation
          positive regulation of protein autophosphorylation
          positive regulation of telomere maintenance
          DNA duplex unwinding
          positive regulation of kinase activity
          isotype switching
          neuromuscular process controlling balance
          meiotic cell cycle
          intrinsic apoptotic signaling pathway
          regulation of signal transduction by p53 class mediator
          negative regulation of telomere capping
          damaged DNA binding
          ATP-dependent DNA helicase activity
          protein binding
          transcription factor binding
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0