Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175214.3 | NP_780423.2 | 14 | 3211 | ||
GenBank mRNA | AK032912.1 | - | 7 | 1066 | ||
BC116646.1 | - | 12 | 2050 | 40086396 |