Search
Search

Fggy| Gene Symbol: Fggy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113412.1 | NP_001106883.1 | 6 | 728 | ||
| NM_029347.2 | NP_083623.1 | 6 | 777 | |||
| GenBank mRNA | AK082737.1 | - | 4 | 412 | ||
| AK085534.1 | - | 6 | 739 | |||
| AK138439.1 | - | 6 | 725 | |||
| BC031708.1 | - | 6 | 695 | 4194716 | ||