Search
Search

Calml4| Gene Symbol: Calml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102468.1 | NP_001095938.1 | 4 | 305 | ||
| NM_138304.2 | NP_612177.1 | 5 | 446 | |||
| GenBank mRNA | AK008039.1 | - | 5 | 432 | ||
| AY061807.1 | - | 5 | 446 | |||
| BC013541.1 | - | 5 | 424 | 4241711 | ||
| BC023475.1 | - | 5 | 411 | 5042406 | ||
| BY097052.1 | - | 4 | 314 | |||