Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SEPT2 SIRNA Assays ›
          Gene Symbol:
          SEPT2
          Gene Name:
          septin 2
          Gene Aliases:
          DIFF6, NEDD-5, NEDD5, Pnutl3, hNedd5
          Chromosome Location:
          Chr.2: 241315187 - 241354027 on Build GRCh38
          UniGene ID:
          Hs.721234
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s9419

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SEPT2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008491.2 NP_001008491.1 5 696
          NM_001008492.2 NP_001008492.1 5 378
          NM_001282972.1 NP_001269901.1 5 427
          NM_001282973.1 NP_001269902.1 7 621
          NM_001321029.1 NP_001307958.1 5 412
          NM_001321030.1 NP_001307959.1 5 410
          NM_001321031.1 NP_001307960.1 6 513
          NM_001321032.1 NP_001307961.1 4 293
          NM_001321033.1 NP_001307962.1 6 529
          NM_001321034.1 NP_001307963.1 6 528
          NM_001321035.1 NP_001307964.1 5 394
          NM_004404.4 NP_004395.1 4 293
          NM_006155.2 NP_006146.1 4 595
          XM_011511237.1 XP_011509539.1 4 243
          XM_011511238.1 XP_011509540.1 3 127
          XM_011511240.1 XP_011509542.1 3 120
          XM_017004189.1 XP_016859678.1 6 488
          XM_017004190.1 XP_016859679.1 6 423
          XM_017004191.1 XP_016859680.1 6 488
          XM_017004192.1 XP_016859681.1 6 488
          XM_017004193.1 XP_016859682.1 4 293
          XM_017004194.1 XP_016859683.1 6 443
          XM_017004195.1 XP_016859684.1 6 647
          XM_017004196.1 XP_016859685.1 6 529
          XM_017004197.1 XP_016859686.1 5 344
          XM_017004198.1 XP_016859687.1 5 406
          XM_017004199.1 XP_016859688.1 6 445
          XM_017004200.1 XP_016859689.1 5 408
          XM_017004201.1 XP_016859690.1 5 530
          XM_017004202.1 XP_016859691.1 4 512
          XM_017004203.1 XP_016859692.1 5 336
          XM_017004204.1 XP_016859693.1 4 251
          XM_017004205.1 XP_016859694.1 6 443
          XM_017004206.1 XP_016859695.1 5 344
          XM_017004207.1 XP_016859696.1 2 145
          XM_017004208.1 XP_016859697.1 5 339
          XM_017004209.1 XP_016859698.1 4 226
          XM_017004210.1 XP_016859699.1 4 270
          XM_017004211.1 XP_016859700.1 3 123
          XM_017004212.1 XP_016859701.1 3 391
          XM_017004213.1 XP_016859702.1 6 488
          GenBank mRNA AF038404.1 - 4 232
          AK294563.1 - 5 335
          BC014455.1 - 5 575 4906575
          BC033559.1 - 7 599 4821654
          BG721140.1 - 4 293
          BI561073.1 - 4 509
          BX648000.1 - 5 312
          CN282712.1 - 4 326
          D63878.1 - 4 412
          DA444624.1 - 6 452
          DA544741.1 - 5 397
          DB091579.1 - 4 293
          DB095611.1 - 4 529
          DB277926.1 - 6 429
          HY043466.1 - 5 400

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of L-glutamate transport
          mitotic nuclear division
          smoothened signaling pathway
          neuron projection development
          regulation of protein localization
          cilium assembly
          regulation of catalytic activity
          cell division
          cell-cell adhesion
          protein binding
          GTP binding
          enzyme regulator activity
          protein complex scaffold
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0