Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDXDC1 SIRNA Assays ›
          Gene Symbol:
          Pdxdc1
          Gene Name:
          pyridoxal-dependent decarboxylase domain containing 1
          Gene Aliases:
          2210010A19Rik, AA415817, Kiaa0251-hp
          Chromosome Location:
          Chr.16: 13833161 - 13903145 on Build GRCm38
          UniGene ID:
          Mm.171484
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s97005

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pdxdc1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039533.2 NP_001034622.1 17 1649
          NM_001291017.1 NP_001277946.1 17 1649
          NM_053181.3 NP_444411.2 17 1649
          GenBank mRNA AK004611.1 - 17 1526
          AK030613.1 - 17 1480
          AK033107.1 - 17 1480
          AK054329.1 - 17 1632
          AK075829.1 - 10 772
          AK163696.1 - 17 1520
          AK165587.1 - 17 1484
          AK178472.1 - 2 118
          AK207349.1 - 2 152
          AK209827.1 - 2 152
          AK210410.1 - 2 152
          BC005541.1 - 17 1467 3590316

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          carboxylic acid metabolic process
          catalytic activity
          phosphoenolpyruvate carboxykinase activity
          gamma-glutamyl carboxylase activity
          3-octaprenyl-4-hydroxybenzoate carboxy-lyase activity
          lyase activity
          carboxy-lyase activity
          2-hydroxy-3-carboxy-6-oxo-7-methylocta-2,4-dienoate decarboxylase activity
          bis(4-chlorophenyl)acetate decarboxylase activity
          3,5-dibromo-4-hydroxybenzoate decarboxylase activity
          2-hydroxyisobutyrate decarboxylase activity
          2-hydroxy-2-methyl-1,3-dicarbonate decarboxylase activity
          pyridoxal phosphate binding
          2-hydroxyisophthalate decarboxylase activity
          dimethylmalonate decarboxylase activity
          2,4,4-trimethyl-3-oxopentanoate decarboxylase activity
          4,4-dimethyl-3-oxopentanoate decarboxylase activity
          2,3,6-trihydroxyisonicotinate decarboxylase activity
          phenanthrene-4,5-dicarboxylate decarboxylase activity
          pyrrole-2-carboxylate decarboxylase activity
          terephthalate decarboxylase activity
          malonate semialdehyde decarboxylase activity
          5-amino-4-imidazole carboxylate lyase activity
          2-keto-4-methylthiobutyrate aminotransferase activity
          2-oxo-4-hydroxy-4-carboxy-5-ureidoimidazoline decarboxylase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0