Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRCAM SIRNA Assays ›
Gene Symbol:
NRCAM
Gene Name:
neuronal cell adhesion molecule
Gene Aliases:
-
Chromosome Location:
Chr.7: 108147626 - 108456442 on Build GRCh38
UniGene ID:
Hs.21422
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s9714
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRCAM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037132.2
NP_001032209.1
30
3861
NM_001193582.1
NP_001180511.1
31
3962
NM_001193583.1
NP_001180512.1
30
3905
NM_001193584.1
NP_001180513.1
29
3869
NM_005010.4
NP_005001.3
28
3878
XM_005250373.2
XP_005250430.1
33
4241
XM_005250383.2
XP_005250440.1
31
3971
XM_005250385.2
XP_005250442.1
30
3926
XM_006716003.1
XP_006716066.1
34
4253
XM_006716007.1
XP_006716070.1
33
4217
XM_006716012.1
XP_006716075.1
32
3974
XM_006716014.1
XP_006716077.1
31
3938
XM_011516253.1
XP_011514555.1
34
4262
XM_011516255.1
XP_011514557.1
33
4017
XM_011516256.1
XP_011514558.1
33
4250
XM_011516257.1
XP_011514559.1
33
4244
XM_011516258.1
XP_011514560.1
32
3999
XM_011516259.1
XP_011514561.1
33
4232
XM_011516261.1
XP_011514563.1
32
4214
XM_011516262.1
XP_011514564.1
33
4205
XM_011516267.1
XP_011514569.1
32
3983
XM_011516268.2
XP_011514570.1
32
4077
XM_011516269.1
XP_011514571.1
31
3947
XM_011516270.1
XP_011514572.1
30
3935
XM_011516271.1
XP_011514573.1
28
3887
XM_017012236.1
XP_016867725.1
33
4223
XM_017012237.1
XP_016867726.1
32
4205
XM_017012238.1
XP_016867727.1
32
4193
XM_017012239.1
XP_016867728.1
32
4187
XM_017012240.1
XP_016867729.1
32
4184
XM_017012241.1
XP_016867730.1
32
3920
XM_017012242.1
XP_016867731.1
30
3872
XM_017012243.1
XP_016867732.1
33
4100
XM_017012244.1
XP_016867733.1
32
4097
XM_017012245.1
XP_016867734.1
32
4088
XM_017012246.1
XP_016867735.1
31
3944
XM_017012247.1
XP_016867736.1
31
3917
XM_017012248.1
XP_016867737.1
30
3917
XM_017012249.1
XP_016867738.1
30
3914
XM_017012250.1
XP_016867739.1
30
3650
XM_017012251.1
XP_016867740.1
30
3908
XM_017012252.1
XP_016867741.1
29
3905
XM_017012253.1
XP_016867742.1
28
3607
XM_017012254.1
XP_016867743.1
29
3896
XM_017012255.1
XP_016867744.1
30
3951
XM_017012256.1
XP_016867745.1
27
3595
XM_017012257.1
XP_016867746.1
29
3660
XM_017012258.1
XP_016867747.1
29
3679
XM_017012259.1
XP_016867748.1
27
3624
GenBank mRNA
AB002341.1
-
29
3797
AJ001057.1
-
30
3740
AK092330.1
-
3
792
AK127035.1
-
1
2032
AK294195.1
-
4
411
AY528240.1
-
3
388
BC114570.1
-
29
3793
40083055
BC115736.1
-
30
3829
40083062
BX538010.1
-
28
3823
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
neuron migration
axonogenesis
axonal fasciculation
synapse assembly
central nervous system development
protein localization
single organismal cell-cell adhesion
neuronal action potential propagation
regulation of axon extension
retinal ganglion cell axon guidance
heterotypic cell-cell adhesion
clustering of voltage-gated sodium channels
positive regulation of neuron differentiation
protein binding
ankyrin binding
protein binding involved in heterotypic cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0