Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NUCB2 SIRNA Assays ›
Gene Symbol:
NUCB2
Gene Name:
nucleobindin 2
Gene Aliases:
HEL-S-109, NEFA
Chromosome Location:
Chr.11: 17260340 - 17331881 on Build GRCh38
UniGene ID:
Hs.654599
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s9773
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
392.00
Your Price
Online offer:
380.65
380.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NUCB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005013.2
NP_005004.1
5, 6
619
XM_017017804.1
XP_016873293.1
7, 8
854
XM_017017805.1
XP_016873294.1
6, 7
770
XM_017017806.1
XP_016873295.1
6, 7
713
XM_017017807.1
XP_016873296.1
6, 7
724
XM_017017808.1
XP_016873297.1
5, 6
658
XM_017017809.1
XP_016873298.1
5, 6
655
XM_017017810.1
XP_016873299.1
7, 8
1125
XM_017017811.1
XP_016873300.1
7, 8
1034
XM_017017812.1
XP_016873301.1
5, 6
619
XM_017017813.1
XP_016873302.1
6, 7
718
XM_017017814.1
XP_016873303.1
6, 7
711
XM_017017815.1
XP_016873304.1
8, 9
1118
XM_017017816.1
XP_016873305.1
6, 7
888
XM_017017817.1
XP_016873306.1
6, 7
813
XM_017017818.1
XP_016873307.1
4, 5
464
XM_017017819.1
XP_016873308.1
4, 5
409
XM_017017820.1
XP_016873309.1
7, 8
854
XM_017017821.1
XP_016873310.1
7, 8
850
XM_017017822.1
XP_016873311.1
5, 6
622
XM_017017823.1
XP_016873312.1
6, 7
726
XM_017017824.1
XP_016873313.1
6, 7
724
XM_017017825.1
XP_016873314.1
5, 6
642
XM_017017826.1
XP_016873315.1
5, 6
655
XM_017017827.1
XP_016873316.1
7, 8
1081
XM_017017828.1
XP_016873317.1
6, 7
711
XM_017017829.1
XP_016873318.1
4, 5
487
GenBank mRNA
AB231311.1
-
3, 4
374
AF052642.1
-
3, 4
374
AF052643.1
-
3, 4
374
AF052644.1
-
3, 4
374
AF450266.1
-
5, 6
588
AK093209.1
-
11, 12
1615
AK097398.1
-
7, 8
1035
AK128739.1
-
9, 10
2286
AK291507.1
-
5, 6
649
FJ224327.1
-
5, 6
593
KM197170.1
-
1, 2
19
X76732.1
-
5, 6
593
Back To Top
More Information
Panther Classification:
Molecular Function -
calmodulin-related
Gene Ontology Categories:
Process(es)
DNA binding
calcium ion binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline