Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TXNDC5 SIRNA Assays ›
          Gene Symbol:
          Txndc5
          Gene Name:
          thioredoxin domain containing 5
          Gene Aliases:
          AL022641, ERp46, PC-TRP
          Chromosome Location:
          Chr.13: 38500266 - 38528824 on Build GRCm38
          UniGene ID:
          Mm.28622
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s98418

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Txndc5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001289598.1 NP_001276527.1 4, 5 635
          NM_001289599.1 NP_001276528.1 4, 5 602
          NM_145367.4 NP_663342.3 4, 5 659
          GenBank mRNA AK136046.1 - 4, 5 547
          AK142049.1 - 4, 5 645
          AK150781.1 - 4, 5 643
          AK169680.1 - 4, 5 626
          AK224911.1 - 3, 4 312
          AK224913.1 - 4, 5 646
          AK224922.1 - 4, 5 604
          AK224923.1 - 4, 5 626
          AK224924.1 - 4, 5 634
          AK224926.1 - 4, 5 604
          AK224927.1 - 4, 5 655
          AK224930.1 - 4, 5 607
          AK224933.1 - 4, 5 596
          AK224934.1 - 4, 5 650
          AK224936.1 - 4, 5 649
          AK224937.1 - 4, 5 648
          AK224938.1 - 4, 5 643
          AK224939.1 - 4, 5 636
          AK224943.1 - 4, 5 607
          AK224945.1 - 4, 5 643
          AK224946.1 - 4, 5 646
          AK224947.1 - 4, 5 633
          AK224948.1 - 4, 5 607
          AK224949.1 - 4, 5 596
          AK224950.1 - 4, 5 596
          AK224951.1 - 4, 5 654
          AK224952.1 - 4, 5 622
          AK224956.1 - 4, 5 566
          AK224957.1 - 4, 5 612
          AK224958.1 - 4, 5 612
          AK224960.1 - 4, 5 654
          AK224962.1 - 4, 5 614
          AK224963.1 - 4, 5 655
          AK224964.1 - 4, 5 655
          AK224965.1 - 4, 5 631
          AK224966.1 - 4, 5 631
          AK224968.1 - 2, 3 189
          AK224969.1 - 2, 3 189
          AK224970.1 - 4, 5 596
          AK224971.1 - 4, 5 656
          AK224972.1 - 4, 5 646
          AK224973.1 - 4, 5 596
          AK224974.1 - 4, 5 599
          AK224976.1 - 4, 5 629
          AK224977.1 - 4, 5 643
          AK224978.1 - 4, 5 643
          AK224979.1 - 4, 5 630
          AK224981.1 - 4, 5 600
          AK224982.1 - 4, 5 655
          AK224983.1 - 4, 5 644
          AK224984.1 - 4, 5 609
          AK224985.1 - 4, 5 613
          AK224986.1 - 4, 5 657
          AK224988.1 - 4, 5 633
          AK224989.1 - 4, 5 650
          AK224990.1 - 4, 5 645
          AK224991.1 - 4, 5 640
          AK224992.1 - 4, 5 640
          AK224993.1 - 4, 5 640
          AK224994.1 - 4, 5 648
          AK224995.1 - 4, 5 611
          AK224996.1 - 4, 5 582
          AK224997.1 - 4, 5 607
          AK224998.1 - 4, 5 605
          AK224999.1 - 4, 5 627
          AK225004.1 - 4, 5 648
          AK225005.1 - 4, 5 626
          AK225007.1 - 4, 5 648
          AK225010.1 - 4, 5 648
          AK225011.1 - 4, 5 648
          AK225012.1 - 4, 5 417
          AK225013.1 - 4, 5 417
          AK225014.1 - 4, 5 626
          AK225015.1 - 4, 5 417
          AK225016.1 - 4, 5 556
          AK225017.1 - 4, 5 604
          AK225019.1 - 4, 5 636
          AK225020.1 - 4, 5 587
          AK225021.1 - 4, 5 587
          AK225022.1 - 4, 5 435
          AK225023.1 - 4, 5 661
          AY243534.1 - 4, 5 622
          AY548112.1 - 4, 5 583
          BC016252.1 - 4, 5 626 4911455
          BC024505.1 - 4, 5 571 4983496
          BC036155.1 - 3, 4 245
          BC046789.1 - 4, 5 585 5716897
          BP765107.1 - 4, 5 649

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001271330, FM067602.

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          response to endoplasmic reticulum stress
          apoptotic cell clearance
          cell redox homeostasis
          protein disulfide isomerase activity
          isomerase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7wptj:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0