Search
Search

Rars2| Gene Symbol: Rars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181406.3 | NP_852071.2 | 5 | 366 | ||
| GenBank mRNA | AK075721.1 | - | 5 | 347 | ||
| AK156182.1 | - | 5 | 344 | |||
| AK166986.1 | - | 5 | 882 | |||
| BC024878.1 | - | 5 | 346 | 3989666 | ||
| BU961647.1 | - | 368 | ||||