Search
All EVOS imaging systems come with onboard software that allows you to view, capture, and process images quickly and easily. You can:
For enhanced cell culture monitoring, the EVOS M3000 Imaging System offers real-time confluency while the EVOS M5000 Imaging System offers additional versatility with cell counting and transfection efficiency applications.
|
||
Cell confluency application |
Real-time confluency |
|
Cell counting application |
- |
|
Transfection efficiency application |
- |
|
For more powerful data generation and complex experiments, the EVOS M5000 Imaging System offers stage-tracking intelligence and autofocus, while the EVOS M7000 Imaging System offers pinpoint operational control and powerful image processing tools such as Z-stacking, image tiling and stitching, and time-lapse movies.
The EVOS S1000 Spatial Imaging System software resolves overlapping emission spectra, eliminating bleed-through and improving data quality for subsequent analysis. Spectral unmixing metrics reports provide qualitative and quantitative insights to assess the quality of the unmixing. Access an exclusive demonstration of the EVOS S1000 operating software.
|
|||
Cell counting application
|
|
|
- |
Stage-tracking intelligence |
|
- |
- |
Z-stacking |
|
|
- |
2D deconvolution |
- |
|
- |
Image tiling and stitching |
- |
|
|
Automated spectral unmixing |
- |
- |
|
Periscope live navigation |
- |
- |
|
Low photobleaching in live mode |
- |
- |
|
EVOS systems are designed with the user in mind, helping make even the most complicated workflows simple to perform. The user-interface shown below represents the typical EVOS system experience.
All these tools operate intuitively with minimal training. We offer onboard SmartStart orientation to provide further assistance as needed.
For advanced image processing and data generation, the optional Celleste 6 Image Analysis Software offers additional tools for classification, segmentation, and analysis of complex images using preconfigured templates with a wizard-based workflow to help simplify batch analysis. Optional modules for deconvolution, 3D visualization, and 3D analysis allow you to customize the capabilities according to your needs.
While we offer advanced software capabilities, all EVOS imaging systems are also designed to enable easy data export for analysis on any software platform, empowering you with the flexibility to discover however you need.