Search
I am having problems with...
![]() | You cannot amplify a SNP. | |
![]() | Not using ROX or not designating it as a passive reference dye may cause trailing clusters. | |
![]() | You have diffuse clustering in your SNP analysis. | |
![]() | Failure of autocalling of SNP results is usually due to outlier wells or software problems. | |
![]() | Normal Allelic Discrimination Plot Example Your cluster plot should look something like this. |
For Research Use Only. Not for use in diagnostic procedures.