Search
Join the conversation
Primary XPS region: Ra4f
Overlapping regions: MgKLL
Binding energies of common chemical states:
| Chemical state | Binding energy Ra4f7/2 |
|---|---|
| Element | 299 eV |
Experimental information
N/A
About this element
Symbol: Ra
Date of discovery: 1898
Name origin: Latin radius
Appearance: silvery white metallic
Discoverer: Marie and Pierre Curie
Obtained from: uranium-bearing ores
Melting point: 700 K
Boiling point: 1737 K
Density[kg/m3]: 5.0
Molar volume: 41.09 × 10-6 m3/mol
Protons/Electrons: 88
Neutrons: 138
Shell structure: 2,8,18,32,18,8,2
Electron configuration: [Rn]7s2
Oxidation state: 2
Crystal structure: cubic
An intensely radioactive element, radium is an alkaline earth metal that is found in trace amounts in uranium ores. Its most stable isotope, 226Ra, has a half-life of 1,602 years and decays into radon gas. Radium emits alpha, beta, and gamma rays, and when mixed with beryllium produces neutrons. Inhalation, injection, or body exposure to radium can cause cancer and other body disorders. In the early 20th century, radium was used to paint watch dials and hands due to its luminescent qualities. The dial painters, mostly women in their early twenties, were some of the first victims of radioactive poisoning.
Products
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.