SQUAD workflows
Simultaneous Quantitation and Discovery ( SQUAD) analysis seamlessly combines discovery and targeted workflows into a single experiment, offering a more comprehensive and versatile solution. Leveraging Thermo Scientific mass spectrometers, SQUAD workflows are enhanced by the unique hardware and software solutions of these platforms.
To enhance depth of discovery analysis, the Thermo Scientific AcquireX workflow optimizes the interrogation of relevant compounds by MS2, leading to greater coverage and more confident annotation when compared to traditional data-dependent analysis.
For robust and precise quantitative measurements with varying sensitivity to meet all application needs, Thermo Scientific mass spectrometers contain Orbitrap, Ion-Trap, and Astral detectors. Additionally, these detectors can operate in parallel configurations enabling researchers to collect all necessary metabolomics data efficiently.
Orbitrap Exploris MS workflows for SQUAD analysis
For comprehensive quantitation and untargeted discovery data in a single injection, SQUAD analysis is conducted on Thermo Scientific Orbitrap Exploris Mass Spectrometers, which combine quadrupole and Orbitrap technologies. Full scan MS1 profiling with rapid polarity switching, at 60K and 120K resolution, provides sufficient points across the peak, complementing high-throughput methods.
Additionally, the AcquireX workflow for intelligent data acquisition is applied to study pool samples and maximizes the depth of metabolite identification that can be achieved in comparison to traditional data dependent MS2.
Three step SQUAD workflow
In the SQUAD workflow, sample preparation includes calibration curves and internal standards where appropriate and available for the targeted analysis portion.
Full scan MS1 profiling with polarity switching is used on all samples and AcquireX iterative data-dependent analysis is performed on the study pools. Data is then processed in two distinct ways:
  • Targeted analysis of known compounds of interest using MzVault library and Thermo Scientific TraceFinder Software
  • Discovery differential analysis and unknown annotation using Thermo Scientific Compound Discoverer Software and Mzcloud online library
Click image to enlarge
Orbitrap Tribrid MS workflows for SQUAD analysis
For fully parallelized acquisition of both targeted and untargeted data, Orbitrap Tribrid Mass Spectrometers combine quadrupole, Orbitrap and ion trap technologies.
During SQUAD analysis on Orbitrap Tribrid Mass Spectrometers, full scan MS1 profiling is performed in the Orbitrap for untargeted discovery data acquisition while known metabolites of interest are detected using targeted-MS2 experiments with sensitive ion-trap detection.
Click image to enlarge
Targeted MS2 ion-trap methods
Compared to full scan Orbitrap quantitation, targeted MS2 ion-trap methods often exhibit an additional order of magnitude linearity at the low end of the curve. At a concentration of 5 femtomoles on the column, sufficient scans of the spiked isotope-labeled phenylalanine in NIST SRM 1950 plasma were observed, achieving a linear dynamic range of six orders of magnitude.
Click image to enlarge
Linear ion trap mass analyzer for accurate and sensitive targeted analysis. A sufficient number of scans of the spiked isotope-labeled phenylalanine in NIST SRM 1950 plasma were observed with 5 femto mole on the column.
Click image to enlarge
Absolute quantitation results for isotope-labeled phenylalanine spike in NIST SRM 1950 plasma reference standard.
AcquireX workflow for intelligent data acquisition
Additionally, the AcquireX workflow for intelligent data acquisition is applied to study pool samples, maximizing the range of relevant compounds interrogated by MS2. This approach increases compound identifications and reduces redundant fragmentations commonly associated with traditional data-dependent methods.
What are customers saying about SQUAD Analysis on the IQ-X
"Quantification is the next frontier in metabolomics and lipidomics. Optimal use of targeted ion-trap MS/MS time in the SQUAD approach is key for getting reliable peak intensities for low abundant compounds without compromising the untargeted full scan orbitrap discovery analysis. This advance is critically needed in the field."
Oliver Fiehn
Department of Molecular and Cellular Biology, University of California Davis
SQUAD Analysis: Maximizing metabolomics information obtained by mass spectrometry
SQUAD analysis enhances our understanding of molecular relationships within biological systems and on biological samples.
“The ability of conducting hypothesis-generating (non-targeted) and hypothesis-testing (targeted) analyses in the same LC-MS experiment is a game changer, saving us precious time and resources. Quantitative targeted ion trap data is highly robust across batches, minimizing any experimental drifts or artifacts.”
Facundo M. Fernández, PhD.
Regents’ Professor and Vasser-Woolley Chair in Bioanalytical Chemistry, School of Chemistry and Biochemistry & Petit Institute for Bioengineering & Bioscience (IBB), Georgia Institute of Technology
Orbitrap Astral MS workflows for SQUAD analysis
The Orbitrap Astral Mass Spectrometer combines quadrupole, Orbitrap, and the innovative Astral mass analyzer for high-throughput SQUAD capabilities with exceptional speed, sensitivity, and annotations. During SQUAD analysis, the Orbitrap performs full scan MS1 profiling, while the Astral detector achieves comprehensive metabolome MS2 coverage.
A single metabolomics injection on the Orbitrap Astral MS provides over 90% MS2 coverage of all compounds in the sample. This rapid performance eliminates the need to investigate only pool samples for identification, enabling in-depth MS2 analysis of all samples in your study.
On an Orbitrap Astral MS, full scan detection is done in the orbitrap while either data-dependent or targeted MS2 is performed in the Astral mass analyzer. This allows for both discovery and targeted to be collected simultaneously while exploiting the speed and sensitivity of the HRAM Astral.
A 15-minute gradient can be reduced to increase throughput by exploiting the speed of the Orbitrap Astral MS.
The number of Orbitrap MS1 scans across the plasma phenylalanine peak of 15- and 5-min LC gradients remain high, while the percentage of compound fragmentation achieved in data-dependent MS2 analysis (DDA) remains above 90%.
Click image to enlarge
Click image to enlarge
What are customers saying about SQUAD analysis on the Orbitrap Astral MS
“The capability to generate MS/MS data for nearly all compounds in a single, short run on an Orbitrap Astral MS is a huge step forward in metabolomics. It will be particularly important for characterizing exposure chemicals, such as drugs and certain food components.”
Gary Patti, Ph.D.
Departments of Chemistry, Genetics, Medicine, Washington University in St. Louis
Leading the way in metabolomics
Unlock the potential of metabolomics to advance human health. For extensive and detailed analysis of the metabolome, experience exceptional analyte separation using innovative gas, liquid, and ion chromatography systems combined with a Thermo Scientific Orbitrap mass spectrometer.
The LC-MS are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our Products have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in inaccurate results and, more seriously, health and safety risks. They are designated for Research Use Only Applications.