Media manufacturing services
Fast pilot-scale media manufacturing services
Accelerate your media development and scale-up with small batch, custom media manufacturing solutions for research and process development from Thermo Fisher Scientific.
Utilizing our dedicated non-GMP labs and cGMP-quality raw materials and processes, our team can provide media manufacturing expertise in liquids, dry powder media (DPM), and our proprietary Advanced Granulation Technology (AGT) media format.
Test the manufacturability and scalability of your formulation or modify existing formulations by adding novel components or removing components not driving cell culture performance. In preparation for cGMP scale up, our team can convert your formulation to a new media format to better suit your scale-up needs. We can also provide details to help you validate us as a secondary supply source.
Our experienced team will help ensure your formulation is consistent and scalable, streamlining your transfer to our cGMP manufacturing facility when you are ready.
Our Gibco Rapid Prototyping Service* also features:
  • cGMP-quality raw materials and processes for easy transfer to cGMP scale
  • Liquid, DPM, and AGT formats—available with batches up to 600 L or 30 kg (DPM)
  • Rapid lead times—standard orders are typically shipped within 10–20 working days*
  • Both pin mill and FitzMill™ capabilities
  • Standard testing includes pH, osmolality, appearance, sterility, and solubility
*While our Gibco Rapid Prototyping service can use animal origin-free (AOF) raw materials, manufacturing does not take place in an animal origin-free (AOF) facility due to lack of dedicated equipment and segregation.
Media format availability:
Media format
Turnaround time*
Volume
Packaging
Liquid media (1X) and liquid
concentrate
2 weeks
1 to 600 L
Standard bottles and
Thermo Scientific
BioProcess Containers (BPCs)
Dry powder media (DPM)
2 weeks
1 to 30 kg batches
Standard packaging
Advanced Granulation Technology (AGT) media
4 weeks
1 to 8 kg batches
Standard packaging
*Complex formulations, novel raw materials, and custom packaging may increase manufacturing times.
Webinar: Streamlining cell culture media manufacturing from rapid prototyping through cGMP
Collaborating for operational success
Learn about our global cell culture media manufacturing network’s capabilities and equivalency strategies to help ensure on-time, in-full order fulfillment, support reducing risk and lead times, and to leverage the consistent, reliable quality you depend on.
Do you require media manufacturing services for your own proprietary formulation?
Contact our team today to discover how our services can streamline your media manufacturing needs.
Contact us
Exterior of Gibco cell culture media manufacturing facility Grand Island New York
Discover our media manufacturing capabilities by taking a virtual tour through our facilities.
Cell culture scientist performing media manufacturing analytics
Explore our curated media manufacturing content library for additional resources.
Bioprocessing resources
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing: