The Thermo Scientific ExSolve wafer TEM prep (WTP) DualBeam (FIB-SEM) dramatically reduces the cost and increases the speed of sample preparation, providing semiconductor and data storage manufacturers with quick and easy access to the data they need to verify and monitor process performance. The ExSolve DualBeam can prepare site-specific TEM lamella, sampling many sites per wafer in a fully-automated process inside the fab, giving semiconductor manufacturers much more information than conventional approaches, while at the same time reducing the capital cost of sample preparation by up to 70 percent.
The ExSolve WTP DualBeam system is an automated, high-throughput sample preparation system that can prepare site-specific, 20 nm thick lamellae on whole wafers up to 300 mm in diameter. It is part of a fast, complete workflow that includes Thermo Scientific TEMLink, and the Thermo Scientific Metrios TEM. The ExSolve DualBeam includes FOUP handling and is designed to be located in the fab near the manufacturing line.
The ExSolve WTP DualBeam workflow addresses the needs of customers that require automated, high-throughput sampling at advanced technology nodes. It complements the capabilities of the Thermo Scientific Helios NanoLab DualBeam 1200AT, which provides more flexible, operator-directed, sample preparation methods, along with  additional capabilities such as high-resolution scanning electron microscopy (SEM) imaging and analysis.
ExSolve TEM Lamella
Applications
Semiconductor-RD_1920x1080
Semiconductor research and development
Innovation starts with research and development. Learn more about solutions to help you understand innovative structures and materials at the atomic level.
Semi-Metrology_1920x1080
Semiconductor metrology
Manufacturing today’s complex semiconductors requires exact process controls. Learn more about advanced metrology and analysis solutions to accelerate yield learnings.
Semiconductor Failure Analysis
Semiconductor Failure Analysis
Complex semiconductor device structures result in more places for defects to hide. Learn more about failure analysis solutions to isolate, analyze, and repair defects.
Semi-Materials-Character_1920x1080
Semiconductor materials characterization
Many factors impact yield, performance, and reliability. Learn more about solutions to characterize physical, structural, and chemical properties.
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Komodo Tabs
Techniques
Sample Preparation of Semiconductor Devices
Thermo Scientific DualBeam systems provide accurate TEM sample preparation for atomic-scale analysis of semiconductor devices. Automation and advanced machine learning technologies produce high-quality samples, at the correct location, and a low cost per sample.
Learn more ›
TEM Metrology
Advanced and automated TEM metrology routines deliver significantly greater precision than manual methods. This allows users to generate large amounts of statistically relevant data, with sub-angstrom-level specificity, that is free of operator bias.
Learn more ›
Sample Preparation of Semiconductor Devices
Thermo Scientific DualBeam systems provide accurate TEM sample preparation for atomic-scale analysis of semiconductor devices. Automation and advanced machine learning technologies produce high-quality samples, at the correct location, and a low cost per sample.
Learn more ›
TEM Metrology
Advanced and automated TEM metrology routines deliver significantly greater precision than manual methods. This allows users to generate large amounts of statistically relevant data, with sub-angstrom-level specificity, that is free of operator bias.
Learn more ›
Contact us
Electron microscopy services for
semiconductors