Search
We manufactured these Thermo Scientific GC columns using proprietary polymer and surface treatments to overcome problems traditionally associated with high-polarity columns and difficult-to-analyze separations.
Column | Recommended Application |
|---|---|
| TraceGOLD™ TG-WaxMS GC Columns | Analyze FAMEs using these columns with a polar-deactivated surface that tightly binds polymer, providing thermal stability and a resistivity to damage from oxidation and from strongly acidic or basic volatiles better than silicone solid phases. |
| TraceGOLD™ TG-WaxMS GC Columns with SafeGuard | Produce consistent results with this low-bleed, stable column integrated with Safeguard to extend column life without compromising performance. |
| TraceGOLD™ TG-WaxMT Metal GC Columns | Ensure excellent column-to-column reproducibility with these thermally stable columns that are resistive to damage from oxidation and from strongly acidic or basic volatiles |
| TraceGOLD™ TG-WaxMS A GC Columns | Identify organic acids with high performance, sensitivity and reproducibility while preventing oxidative damage with these low-bleed columns |
| TraceGOLD™ TG-WaxMS B GC Columns | Identify basic analytes with high performance, sensitivity and reproducibility without derivatization or column priming using these collumns |
| TraceGOLD™ TG-POLAR GC Columns | Use the low bleed, stable columns to produce consistent results while overcoming problems traditionally associated with polar colums |
| TRACE™ TR-Wax GC Columns | Analyze a wide range of polar compounds with these cross-linked and fully deactivated polyethylene-glycol-based columns optimized for all polar compounds to provide excellent peak shape even with short chain alcohols |
| TRACE™ TR-WaxMS GC Columns | For high-polarity separations as part of GC-MS analysis, choose these low-bleed columns manufactured using a proprietary bonding technique that minimizes bleed and extends the maximum operating temperature |
| TRACE™ TR-FFAP GC Columns | For outstanding robustness in difficult-to-analyze separations even at elevated temperatures, choose these low-bleed columns that are quality-tested for acidic compound analysis |
| TRACE™ TR-FAME GC Columns | Use these cyanopropylphenyl-based columns optimized for the separation of cis/trans isomers and FAMEs with various degrees of unsaturation and for any high polarity GC application used with MS detection |
Find interactive tools and calculators, guides to select the right products, equivalent product finders, and get in contact with our consumables technical support team to answer your application questions.
Access the latest interactive version of our Chromatography Consumables & Columns Catalog. Our enhanced online version features easier access to product information, key application notes and brand-new selector tools.