Search
This array of compact, powerful instruments based on the Thermo Scientific™ ARL™ OPTIM'X WDXRF Spectrometer continues a tradition of innovative leadership in wavelength dispersive fluorescence spectroscopy. Each analyzer comes delivered as a turnkey solution preconfigured to characterize with efficiency, precision, repeatability, speed and simplicity of sample preparation materials used in the petroleum, cement or slag industries, or in multi-purpose analytical laboratories.
Package | Application |
|---|---|
| ARL OPTIM'X Sulfur Analyzer | A uniquely compact and reliable WDXRF platform configured with dedicated multichromators for ultra-low sulfur analysis in heavy and light fuels and oils |
| ARL OPTIM'X Cement Analyzer | A uniquely compact and reliable WDXRF platform configured with Thermo Scientific™ SmartGonio™ for analysis of raw meal, clinker and cement |
| ARL OPTIM'X Slag Analyzer | A uniquely compact and reliable WDXRF platform configured with Thermo Scientific™ SmartGonio™ for analysis of slags and pig iron |
• ARL OPTIM'X at SGS Belgium-Antwerp: Where speed of sulfur analysis and ease of use form a perfect solution
• NEDSTAAL combines routine quality control and versatile elemental analysis using one instrument: the Thermo Scientific ARL OPTIM'X WDXRF Spectrometer
To be regularly informed on our latest developments and promotions (on OES, XRF, XRD and related techniques), subscribe to our E-Newsletter MATERIALS ANALYST.