Search
As your work evolves, so do the needs of your lab. Our orbital shaking machines can keep up with your growing needs. Whatever your research requirements, we have a shaker suited to the job.
A shaker table’s drive system defines its performance and durability. We use heavy duty, triple eccentric, and triple counterbalanced drive mechanisms in our shakers. The triple eccentric drive accommodates heavier loads, while the triple counterbalanced drive supports unbalanced loading. Both systems enable uniform, reliable operation over time.
Model Type |
Orbit Sizes |
Temperature Ranges |
Speed Control Ranges |
Open Air Shakers |
0.4 in. (1.2cm) 0.5 in. (1.3cm) 1 in. (2.5cm) |
Ambient Ambient + 5°C to 65°C Ambient |
40-400 rpm 15-500 rpm 15-525 rpm |
CO2 Resistant Shakers |
.118 in. (0.3 cm) .75 in. (1.9 cm) 1 in. (2.5 cm) |
Ambient Ambient Ambient |
30-600 rpm 30-300 rpm 25-525 rpm |
Incubated Shakers |
.75 in (1.9cm) 1 in. (2.54cm) 1 in. (2.54cm) 1 in. (2.54cm) |
Ambient + 10°C to80°C Ambient + 10°C to 60°C Ambient + 5°C to 60°C + 30°C to 60°C |
15-500 rpm 25-400 rpm 25-525 rpm 15-525 rpm |
Refrigerated and Incubated Shakers |
0.75 in. (1.9cm) 1 in. (2.54cm) 1 in. (2.54cm) 1 in. (2.54cm) 1 in. (2.54cm) |
4°C to 80°C 5°C below ambient to 60°C 4°C to 60°C 5°C to 60°C 4°C to 60°C |
15-500 rpm 25-400 rpm 25-525 rpm 15-525 rpm 15-525 rpm |
Our shakers include ergonomic features like bright, intuitive controls and easy-to-reach and easy-to-clean surfaces that are appreciated by busy staff. Quiet operation and a variety of sizes and configurations that help maximize capacity while saving valuable lab space also contribute to a better lab environment.
As your work evolves, so do the needs of your lab. Our orbital shaking machines can keep up with your growing needs. Whatever your research requirements, we have a shaker suited to the job.