Search
Our portfolio of orbital shakers offers a wide selection of formats, features and capabilities that are ideal for nearly any application and lab environment. Product families have interchangeable clamps and fasteners and are organized together in the tables below. Browse the tables to find your next orbital shaker, or use our selection guide to compare models.
|
|
|
Installation |
||
Floor |
Floor |
Bench |
Orbital diameter |
||
1 – 2 in. / 25 – 50 mm |
1 – 2 in. / 25 – 50 mm |
.12 – .75 in. / 3 – 19 mm |
Platform size |
||
35 x 22 in. / 89 x 56 cm |
35 x 22 in. / 89 x 56 cm |
14.1 x 11.8 in. / 36 x 30 cm, or 18 x 18 in. / 46 x 46 cm |
Unit weight |
||
684 lbs. / 301 kg
|
684 lbs. / 301 kg | 29 lbs. / 13.2 kg |
Largest flask size (clamp) |
||
5 L |
5 L |
5 L |
Max capacity of largest flask (using adhesive mats) |
||
8 |
8 |
3 |
| Max load capacity | ||
44.1 lbs. / 20 kg |
44.1 lbs. / 20 kg |
13 lbs. / 5.9 kg |
| Speed range | ||
300 – 350 ± 1 rpm, 25 mm dia. 300 – 300 ± 1 rpm, 50 mm dia. |
300 – 350 ± 1 rpm, 25 mm dia. 300 – 300 ± 1 rpm, 50 mm dia. |
30 - 1,200 ± 1 rpm, 3 mm dia. 30 - 300 ± 1 rpm, 19 mm dia. |
| Temperature range | ||
4 – 60 °C |
4 – 60 °C |
-- |
| Additional features | ||
Temperature control Humidity control CO2 resistant Access port Programmable |
Temperature control Humidity control CO2 control CO2 resistant Access port Programmable |
CO2 resistant |
|
Solaris Incubated and Refrigerated Benchtop Temperature Control Shaker |
|
| Installation | ||
Bench or floor, stackable |
Bench |
Bench |
Orbital diameter |
||
.75 in. / 19 mm |
1 in. / 25.4 mm |
1 in. / 25.4 mm |
Platform size |
||
18 x 18 in. / 46 x 46 cm |
11 x 14 in. / 31 x 36 cm, or 18 x 18 in. / 46 x 46 cm |
12 x 14 in. / 31 x 36 cm, or 18 x 30 in. / 46 x 76 cm, or 36 x 24 in. / 92 x 61 cm |
Unit weight |
||
300 lbs. / 150 kg incubated 430 lbs. / 195 kg refrigerated |
99 – 216 lbs. /45 – 98 kg |
46 – 166 lbs. / 21 – 75 kg |
Largest flask size (clamp) |
||
6 L |
2 L |
6 L |
Max capacity of largest flask (using adhesive mats) |
||
4 |
4 |
8 |
| Max load capacity | ||
35 lbs. / 16 kg |
35 – 71 lbs. / 16 – 32 kg |
55 – 95 lbs. / 25 – 43 kg |
| Speed range | ||
15 – 500 ± 1 rpm |
15 – 525 ± 1 rpm |
15 – 525 ± 1 rpm |
| Temperature range | ||
Ambient + 10 – 80 °C incubated 4 – 80 °C refrigerated |
30 – 60 °C incubated 5 – 60 °C refrigerated |
4- 60 °C |
| Additional features | ||
Temperature control |
Temperature control Access port Programmable |
Programmable |
|
|
| Installation | |
Floor |
Bench |
Orbital diameter |
|
1 in. / 25.4 mm |
1 in. / 25.4 mm |
Platform size |
|
30 x 18 in. / 76 x 46 cm
|
18 x 18 in. / 46 x 46 cm, or 24 x 18 in. / 61 x 46 cm |
Unit weight |
|
550 lbs. / 250 kg incubated 600 lbs. / 273 kg refrigerated |
138 lbs. /62.7 kg |
Largest flask size (clamp) |
|
6 L |
2 L |
Max capacity of largest flask (using adhesive mats) |
|
6 |
4 |
| Max load capacity | |
71 lbs. / 31.2 kg |
71 lbs. / 31.2 kg |
| Speed range | |
25 – 400 ± 1 rpm |
25 – 525 ± 1 rpm |
| Temperature range | |
Ambient + 10 – 60 °C incubated Ambient - 5 – 60 °C refrigerated |
-- |
| Additional features | |
Temperature control Programmable |
CO2 resistant Programmable |